Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5b Antibody (STAT5B/2611), Janelia Fluor™ 646, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP279907JF646

Catalog No. NB012313


Only null left
Add to Cart

Description

Description

STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

STAT5b
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
STAT5B
Recombinant full-length human STAT5b protein (Uniprot: P51692)
0.1 mL
Phospho Specific, Signal Transduction
Human
IgG1 κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
STAT5B/2611
Immunohistochemistry, Immunohistochemistry-Paraffin
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.