Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5b Antibody (STAT5B/2657), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308628AF488
Description
STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for ELISA, Protein ArraySpecifications
STAT5b | |
Monoclonal | |
Alexa Fluor 488 | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
Recombinant full-length human STAT5b protein (Uniprot: P51692) | |
100 μg | |
Signal Transduction | |
6777 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Protein Array | |
STAT5B/2657 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction