Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5b Rabbit anti-Human, Mouse, Rat, Clone: 6P5T0, Novus Biologicals™
SDP

Catalog No. NB166652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166652 20 μg
NB166651 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166652 Supplier Novus Biologicals Supplier No. NBP31577820UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

STAT5b Monoclonal antibody specifically detects STAT5b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, KnockDown

Specifications

Antigen STAT5b
Applications Western Blot, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 6P5T0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 600-787 of human STAT5b (P51692).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6777
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.