Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT6 Chicken anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209260
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9260 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9260 Supplier Fabgennix Inc Supplier No. STAT6101Y
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

STAT6 Polyclonal antibody specifically detects STAT6 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen STAT6
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene STAT6
Gene Accession No. P42226, P52633
Gene Alias 12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT
Host Species Chicken
Immunogen Synthetic peptide taken within amino acid region 655-705 on human Signal transducer and activator of transcription 6 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20852, 362896, 6778
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.