Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STAT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587502
Description
Positive Samples: HeLa, Jurkat, A-431, THP-1; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: AACTCCTGCT AATGGGACTG GGCCAAGTGA GGCCCTGGCC ATGCTACTGC AGGAGACCAC TGGAGAGCTA GAGGCAGCCA AAGCCCTAGT GCTGAAGAGG.
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.
Specifications
STAT6 | |
Polyclonal | |
Unconjugated | |
STAT6 | |
12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT | |
Rabbit | |
Affinity chromatography | |
RUO | |
20852, 362896, 6778 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2.07 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P42226, P52633 | |
STAT6 | |
A synthetic peptide corresponding to a sequence within amino acids 501-600 of human STAT6 (NP_0031443). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction