Learn More
Stat6 (pY641) Mouse anti-Human, PE, Clone: 18/P-STAT6, BD
Mouse Monoclonal Antibody
$355.50 - $1601.00
Specifications
| Antigen | Stat6 (pY641) |
|---|---|
| Clone | 18/P-STAT6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
Description
Interleukin-4 (IL-4), a major immunoregulatory cytokine, is secreted by activated T lymphocytes, basophils, and mast cells and plays an important role in modulating T helper cell lineage development. It induces specific gene expression via the tyrosine phosphorylation of Stat6 at tyrosine 641 (Y641). Stat6, a member of the signal transducers and activators of transcription protein family, mediates signals for IL-4 and, possibly, IL-13. While Stat6 is widely expressed in human tissues, it exhibits elevated expression in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung, and placenta. Following cytokine receptor ligation, Jak kinases are activated and phosphorylate the cytoplasmic tails of the oligomerized receptors. The SH3:SH2 domain of Stat6 associates with tyrosine-phosphorylated IL-4 receptor and the proximal Jak kinase phosphorylates Stat6 at Y641 on the C-terminal side of the SH2 domain. Stat6 is then released from the receptor, dimerizes, and is thought to contact the basal transcription machinery by binding to p300/CBP. Thus, Stat6 mediates the IL-4 signal and is essential for the proper development of adaptive immunity.
Specifications
| Stat6 (pY641) | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified | |
| Signal transducer and activator of transcription 6; IL-4 Stat |
| 18/P-STAT6 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Phosphorylated Human Stat6 (pY641) | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.