Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT6 Rabbit anti-Human, Mouse, APC, Clone: G12, Abnova™

Catalog No. 89335986 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
100 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-986 10 Reactions
89-335-985 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-335-986 Supplier Abnova Corporation Supplier No. MAB19152
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

STAT6 Monoclonal antibody specifically detects STAT6 in Human, Mouse samples. It is validated for Flow Cyt.

Specifications

Antigen STAT6
Applications Flow Cytometry
Classification Monoclonal
Clone G12
Conjugate APC
Description signal transducer and activator of transcription 6, interleukin-4 induced
Gene STAT6
Gene Alias D12S1644, IL-4-STAT, STAT6B, STAT6C
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y641 of human STAT6
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 6778
Target Species Human, Mouse
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.