Learn More
StatLab™ StatCut™ Microtome Blades
GSA_VASDP
Catalog No. p-7227177
Click to view available options
Profile:
High
Low
This item is not returnable. View return policy
This item is not returnable. View return policy
Suitable for both soft and hard tissue, StatCut™ blades are well balanced in durability and sharpness.
  • Blades are coated for non-stick sections and effortless ribboning
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Profile Low
Disposable Single-use
Quantity 50 Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.