Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STATH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542103 Supplier Invitrogen™ Supplier No. PA542103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MKFLVFAFIL ALMVSMIGAD SSEEKFLRRI GRFGYGYGPY QPVPEQPLYP Sequence homology: Human: 100%; Yeast: 90%.

STATH is a salivary protein that stabilizes saliva supersaturated with calcium salts by inhibiting the precipitation of calcium phosphate salts. It also modulates hydroxyapatite crystal formation on the tooth surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STATH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene STATH
Gene Accession No. P02808
Gene Alias STATH; Statherin; STR
Gene Symbols STATH
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human STATH.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6779
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.