Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stathmin 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298752AF488
Description
Stathmin 1 Polyclonal specifically detects Stathmin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Stathmin 1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
C1orf215, chromosome 1 open reading frame 215, Lag, Leukemia-associated phosphoprotein p18, Metablastin, MGC138869, Oncoprotein 18, Op18, OP18MGC138870, phosphoprotein 19, Phosphoprotein p19, pp17, PP19, PR22, prosolin, Protein Pr22, SMNLAP18FLJ32206, stathmin, stathmin 1, stathmin 1/oncoprotein 18, transmembrane protein C1orf215 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Stathmin 1. | |
0.1 mL | |
Breast Cancer, Cancer, Cognition and Behavior, Immunology, Microglia Markers, Neuroscience, Phospho Specific, Signal Transduction | |
3925 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction