Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Stathmin 1 Monoclonal Antibody (SP49)
GREENER_CHOICE

Catalog No. PIMA516400
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516400 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516400 Supplier Invitrogen™ Supplier No. MA516400
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil or breast carcinoma.

Stathmin 1 belongs to the stathmin family of genes. It encodes a ubiquitous cytosolic phosphoprotein proposed to function as an intracellular relay integrating regulatory signals of the cellular environment. Stathmin 1 is involved in the regulation of the microtubule filament system by destabilizing microtubules. It prevents assembly and promotes disassembly of microtubules. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stathmin 1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone SP49
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene Stmn1
Gene Accession No. P13668, P16949, P54227
Gene Alias 19k; C1orf215; I79_017954; Lag; LAP18; leukemia associated phosphoprotein p18; leukemia-associated cytosolic phosphoprotein stathmin; leukemia-associated gene protein; Leukemia-associated phosphoprotein p18; metablastin; Oncoprotein 18; Op18; OTTHUMP00000008529; OTTHUMP00000008531; P18; P19; phosphoprotein 19; phosphoprotein p19; Pig; pp17; Pp18; Pp19; Pr22; Pr22 protein; Prosolin; Protein Pr22; RP1-125I3.5; SMN; Stathmin; stathmin 1; stathmin 1 variant; stathmin 1/oncoprotein 18; Stmn1; STN1; testicular tissue protein Li 189; transmembrane protein C1orf215
Gene Symbols Stmn1
Host Species Rabbit
Immunogen Synthetic peptide derived from the internal region of human stathmin.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16765, 29332, 3925
Target Species Human, Mouse, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.