Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Stathmin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520382
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520382 Supplier Invitrogen™ Supplier No. PA520382
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is EL4 cell lysate. PA5-20382 can be used with blocking peptide PEP-0499.

Stathmin 1 belongs to the stathmin family of genes. It encodes a ubiquitous cytosolic phosphoprotein proposed to function as an intracellular relay integrating regulatory signals of the cellular environment. Stathmin 1 is involved in the regulation of the microtubule filament system by destabilizing microtubules. It prevents assembly and promotes disassembly of microtubules. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stathmin 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Stmn1
Gene Accession No. P16949, P54227
Gene Alias 19k; C1orf215; I79_017954; Lag; LAP18; leukemia associated phosphoprotein p18; leukemia-associated cytosolic phosphoprotein stathmin; leukemia-associated gene protein; Leukemia-associated phosphoprotein p18; metablastin; Oncoprotein 18; Op18; OTTHUMP00000008529; OTTHUMP00000008531; P18; P19; phosphoprotein 19; phosphoprotein p19; Pig; pp17; Pp18; Pp19; Pr22; Pr22 protein; Prosolin; Protein Pr22; RP1-125I3.5; SMN; Stathmin; stathmin 1; stathmin 1 variant; stathmin 1/oncoprotein 18; Stmn1; STN1; testicular tissue protein Li 189; transmembrane protein C1orf215
Gene Symbols Stmn1
Host Species Rabbit
Immunogen A 20 amino acid peptide from near the carboxy terminus of human op18.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16765, 3925
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.