Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stathmin 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | Stathmin 1 |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Stathmin 1 Polyclonal antibody specifically detects Stathmin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Stathmin 1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cognition and Behavior, Immunology, Microglia Markers, Neuroscience, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
3925 | |
IgG | |
Affinity purified |
Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
C1orf215, chromosome 1 open reading frame 215, Lag, Leukemia-associated phosphoprotein p18, Metablastin, MGC138869, Oncoprotein 18, Op18, OP18MGC138870, phosphoprotein 19, Phosphoprotein p19, pp17, PP19, PR22, prosolin, Protein Pr22, SMNLAP18FLJ32206, stathmin, stathmin 1, stathmin 1/oncoprotein 18, transmembrane protein C1orf215 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: AFELILSPRSKESVPEFPLSPPKKKDL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title