Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stathmin-2/STMN2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001403 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001403 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001403 Supplier Novus Biologicals Supplier No. NB1001403
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Stathmin-2/STMN2 Polyclonal specifically detects Stathmin-2/STMN2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Stathmin-2/STMN2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_008960.
Gene Alias FLJ34868, FLJ50995, neuron-specific growth-associated protein, Protein SCG10, SCG10, SCG10neuronal growth-associated protein (silencer element), SCGN10stathmin-2, stathmin-like 2, superior cervical ganglia, neural specific 10, Superior cervical ganglion-10 protein
Gene Symbols STMN2
Host Species Goat
Immunogen Peptide with sequence AKTAMAYKEK-C corresponding to N-Terminus according to NP_008960.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11075
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.