Learn More
Static Solutions™ OHM-Stat™ CT-8900 Combination Tester/Data Logger
Description
Includes
NIST certificate.
Measures the electrical resistance of each foot and wrist strap simultaneously and individually with one push of a button. Offers adjustable limits, exact resistivity readings, unique report generation groups, versatile networking options, and an expandable employee log of 30,000. Made in USA.
- Increases worker productivity, reduces defects, and saves administrative time
- Accepts mag stripe, barcode, Motorola™, HID™, and Casi-Rusco™ proximity ID cards as well as keyboard entry
- Included ESD Protection Test Log software generates e-mail reports to multiple recipients
- Logs are easily imported into spreadsheet/database programs
- Prevents cheating—exception report will inform if worker did not test once or twice per day
- Able to prevent unauthorized entry without proper ID/ESD certification; can be upgraded to open electronically locked doors with pass of test
- Measures from 100 kilohms to 100 megohms, with an option to 1000 megohms
- Tests performed at 19 volts—not competitive 9 volts
Order Info
Compatible with Microsoft™ Windows™ 95, 98, ME, NT, and 2000 operating systems. Can use Oracle™, Access™, or Excel™ databases. Powered by 9 volt battery (IEC6F22) or 12 volt DC adapter.
Compliance
Conforms to ESD-S 20-20, European EN, DIN and military standards. NIST and RoHs compliant. CE listed.
Specifications
Specifications
| Includes | Combination Tester/Data Logger |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.