Learn More
Meridian Bioscience™ StatID PRO™ COVID-19/Flu A and B
Catalog No. 21720022 Shop All Meridian Bioscience Products
Change view
Click to view available options
Type:
COVID Testing Kit
Catalog No. Type
21-720-022 COVID Testing Kit
1 options
Catalog No. 21-720-022 Supplier Meridian Bioscience™ Supplier No. 783025
 Restricted Item
Add to Cart
Add to Cart
Fast and reliable rapid testing to support timely clinical decision-making in patients presenting with respiratory symptoms.
  • One swab delivers antigen results for COVID-19, influenza A, and influenza B.
  • FDA-cleared rapid test (not EUA), providing trusted regulatory approval.
  • Approved for use in patients 2 years of age and older.
  • Provides clear visual results in 15 minutes to support immediate clinical decision-making.
  • Offers workflow flexibility, allowing specimens to be held in the extraction tube for up to 1 hour before testing.
Specifications
CE Marker No
Clia Complexity Waived
Detectable Analytes COVID-19
Type COVID Testing Kit
Disposable Single-use
Sterility Non-sterile
Form Ready-to-Use (RTU)