Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab StatPrep CytoLysis Solution
SDP

Catalog No. p-200007156
Change view
Click to view available options
Size:
30 mL
32 oz.
Catalog No. Size
CY32LYSIS32 32 oz.
CY50LYSIS30 30 mL
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Catalog No. CY32LYSIS32

Supplier: StatLab CY32LYSIS/32

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Cytology specimen preservative.

  • Cytology preservative

Specifications

Type Preservative
Autoclavable Not Autoclavable
Size 32 oz.
Sterility Non-sterile
Reusable No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.