Learn More
HemoCue America StatSpin™ Glass Microhematocrit Tubes
Catalog No. p-178568
Change view
Click to view available options
For Use With (Application):
Anti-coagulated samples
Heparinized for fingersticks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
22-269245 Heparinized for fingersticks
22-269246 Anti-coagulated samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-269245 Supplier HemoCue America Supplier No. HT9H10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
HemoCue America StatSpin Glass Microhematocrit Tubes, For Use With: Heparinized for fingersticks, RH12 HCT rotor, Microhematocrit Capillary Tube, Length: 1.57 in., 4cm, Recommended Applications: Healthcare, Shelf Life: 2 Year(s), Volume: 0.0003 oz., 9uL
Glass tubes are 40mm long.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microhematocrit Capillary Tube
Material Glass
Capacity (Metric) 9 μL
For Use With (Equipment) RH12 HCT Rotor
For Use With (Application) Heparinized for fingersticks
Length (Metric) 4 cm
Length (English) 1.57 in.