Learn More
HemoCue America StatSpin™ VT Multipurpose Veterinary Centrifuge
Description
- Compact, less than 7 in. (17.8cm) of bench space
- Quiet operation
- Separates serum or plasma in 30 to 90 seconds, hematocrit in two minutes, and urine sediment in 45 seconds
- Preprogrammed run cycles
- Membrane touch panel and microprocessor control
- Easy-to-clean, coated steel interior
- Remote power supply for installation flexibility
- Cover interlock system for added safety
- Accepts accessories from previous StatSpin models (StatSpin 1, 2, 3, 4; StatSpin MP, RP, CritSpin™)
Compliance
UL listed, CE marked
Warranty and Services
One-year warranty from date of installation; extended warranties available upon request
Specifications
Specifications
| Max. RCF | 13,700 x g |
| Max. Speed | 15,800 rpm |
| Refrigerated | No |
| Profile (Acceleration/Braking) | Approx. 6 sec./<15 sec. |
| Electrical Requirements | 100/240 V 50/60 Hz |
| Certifications/Compliance | UL listed; CE |
| Height (English) Exterior | 6 in. |
| Height (Metric) Exterior | 15.2 cm |
| Length (English) Exterior | 8.6 in. |
| Length (Metric) Exterior | 21.9 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.