Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Stattic
GREENER_CHOICE

Catalog No. 279810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-981-0 10 mg
27-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-981-0 Supplier Tocris Bioscience™ Supplier No. 2798/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective STAT3 inhibitor

Small molecule inhibitor of STAT3. Inhibits binding of tyrosine-phosphorylated peptide motifs to STAT3 SH2 domain and inhibits STAT3 activation, dimerization and nuclear translocation. Displays selectivity over STAT1, STAT5, c-Myc/Max, Jun/Jun and Lck. Induces apoptosis in STAT3-dependent cancer cell lines.

Chemical Identifiers

CAS 19983-44-9
Molecular Formula C8H5NO4S

Specifications

Quantity 10 mg
Formula Weight 211.19
Purity >99%
Molecular Weight (g/mol) 211.191
Chemical Name or Material Stattic
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.