Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Status™ Covid-19/Flu A & B Rapid Test

Catalog No. 23111399
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-399 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-399 Supplier Germaine™ Laboratories Supplier No. 33225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A rapid, lateral flow immunoassay intended for the in-vitro simultaneous qualitative detection and differentiation of nucleocapsid antigen from SARS-CoV- 2, Influenza A, and Influenza B directly from nasopharyngeal swab specimens. The Status COVID-19/Flu Test is only for use under FDA EUA.

  • Status™ Covid-19/Flu A & B Test has been granted emergency use authorization (EUA) by the FDA for use at the point-of-care setting
  • During the duration of the emergency this test can be performed in a patient care setting that is operating under a CUA Waiver, Certificate of Compliance, or Certificate of Accreditation
  • 25 individually pouched test devices
  • 1 test + 1 specimen = 3 Results
  • Results in 15 minutes
  • Made in USA

Includes

  • Extraction Reagent Capsules (25): 300 μL of phosphate buffer with detergents and preservative, for use with swab specimens
  • Sterile Swabs (25): For specimen collection
  • Positive Control Swab (1): Influenza A, B, and SARS-CoV-2 Antigen (Non-infective
    recombinant nucleocapsid protein)
  • Negative Control Swab (1): Inactivated Group B Streptococcus Antigen (Non-infective)
  • Package Insert/Instructions for Use (1)
  • Quick Reference Instruction (1)

Specifications

Certifications/Compliance FDA EUA Approved
Content And Storage 2° to 30°C (35° to 86°F)
Control Sets Includes Positive/Negative Control Swabs
CE Marker Yes
Detectable Analytes SARS-CoV-2, Influenza A/B Nucleic Acids
Sample Type Nucleocapsid Antigen from SARS-CoV2, Influenza A, and Influenza B
Sensitivity COVID-19: 93.9%, Flu A: 91.4%, Flu B: 87.6%
Disposable Single-use
Includes Rapid Test Devices (25), Sterile Swabs (25), Positive Control Swab (1), Negative Control Swab (1), Package Insert/Instructions for Use (1), Quick Reference Instruction (1)
Quantity 25 Tests
Specificity COVID-19: 100%, Flu A: 95.7%, Flu B: 95.9%
Sterility Non-sterile
Test Time 15 min.
Show More Show Less

The Status COVID-19/Flu test is only for use under FDA EUA. Status™ COVID-19/FLU A&B test has been granted emergency use authorization (EUA) by the FDA for use at the point-of-care setting. During the duration of the emergency this test can be performed in a patient care setting that is operating under a CUA Waiver, Certificate of Compliance or Certificate of Accreditation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.