Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSign Status hCG Test Kit
SDP

Catalog No. p-8410000 Shop All Lifesign LLC Products
Encompass
Click to view available options
Sample Type:
Urine
Urine, Serum
Clia Complexity:
Waived
Waived (Urine)

Catalog No. 23-264-793

Supplier: LifeSign 35135

Only null left
Add to Cart
Add to Cart

Rapid immunoassay test for the detection of human chorionic gonadotropin in urine

Features:

  • Excellent sensitivity, 25mIU/mL, regardless of specimen for early pregnancy detection
  • Clear, definitive result lines are easy to read
  • Simple one-step procedure
  • Results in 3 to 5 minutes
  • CLIA waived for urine

Specifications

Clia Complexity Waived
Detectable Analytes Human chorionic gonadotropin
Format Cassette
Sample Type Urine
Sensitivity 25 mIU/mL
Quantity 35 Tests
Test Time 3 to 5 min.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.