Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSign Status Strep A Test Kit
SDP

Catalog No. p-8410003 Shop All Lifesign LLC Products
Encompass
Click to view available options
Quantity:
25 Tests
30 Tests
Format:
Flip-design Cassette
Strip

Catalog No. 23-264-789

Supplier: LifeSign 34125

Only null left
Add to Cart
Add to Cart

Rapid immunochromatographic assay for the detection of group A streptococcal antigen directly from throat swab specimens

Specifications

Clia Complexity Waived
Detectable Analytes Streptococcal Antigen (Group A)
Format Flip-design Cassette
Sample Type Throat Swab
Sensitivity 96.2%
Type Immunochromatographic Assay
Includes External negative and positive controls
Quantity 25 Tests
Specificity 0.987
Test Time 5 to 10 min.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.