Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAU2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582670
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582670 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582670 Supplier Invitrogen™ Supplier No. PA582670
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQINQMFSVQ LSLGEQTWES EGSSIKKAQQ AVANKALTES TLPKPVQKPP KSNVNNNPGS ITPTVELNGL AMKRGEPAIY RPLDPKPFPN YRANYNFRGM YNQRYHCPVP KIFYVQLTVG NNEFFGEGKT RQAARHNAAM KALQ Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 56%.

Staufen homolog 2 is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48. 5% and 59. 9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAU2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Stau2
Gene Accession No. Q68SB1, Q9NUL3
Gene Alias 39K2; 39K3; Double-stranded RNA-binding protein Staufen homolog 2; r-staufen protein; Stau2; staufen (RNA binding protein) homolog 2 (Drosophila); staufen 2; staufen double-stranded RNA binding protein 2; staufen homolog 2; staufen, RNA binding protein, homolog 2
Gene Symbols Stau2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STAU2. Recombinant protein control fragment (Product #RP-102290).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171500, 27067
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.