Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STAU2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589631
Description
Immunogen sequence: PEYGQGMNPI SRLAQIQQAK KEKEPDYVLL SERGMPRRRE FVMQVKVGNE VATGTGPNKK IAKKNAAEAM LLQLGYKAST NLQDQLEKTG ENKGWSGPKP G; Positive Samples: HeLa, A549, Mouse heart, Rat heart; Cellular Location: Cytoplasm, Endoplasmic reticulum, Nucleus, nucleolus.
Staufen homolog 2 is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48. 5% and 59. 9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
STAU2 | |
Polyclonal | |
Unconjugated | |
Stau2 | |
39K2; 39K3; Double-stranded RNA-binding protein Staufen homolog 2; r-staufen protein; Stau2; staufen (RNA binding protein) homolog 2 (Drosophila); staufen 2; staufen double-stranded RNA binding protein 2; staufen homolog 2; staufen, RNA binding protein, homolog 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
171500, 27067, 29819 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.06 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68SB1, Q8CJ67, Q9NUL3 | |
Stau2 | |
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human STAU2 (NP_0011578521). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction