Learn More
Invitrogen™ STAU2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596245
Description
Immunogen sequence: PEYGQGMNPI SRLAQIQQAK KEKEPDYVLL SERGMPRRRE FVMQVKVGNE VATGTGPNKK IAKKNAAEAM LLQLGYKAST NLQDQLEKTG ENKGWSGPKP G; Positive Samples: Mouse heart, Mouse kidney, Rat heart, Rat brain; Cellular Location: Cytoplasm, Endoplasmic reticulum, Nucleus, nucleolus.
Staufen homolog 2 is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48. 5% and 59. 9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| STAU2 | |
| Polyclonal | |
| Unconjugated | |
| Stau2 | |
| 39K2; 39K3; Double-stranded RNA-binding protein Staufen homolog 2; r-staufen protein; Stau2; staufen (RNA binding protein) homolog 2 (Drosophila); staufen 2; staufen double-stranded RNA binding protein 2; staufen homolog 2; staufen, RNA binding protein, homolog 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171500, 27067, 29819 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q68SB1, Q8CJ67, Q9NUL3 | |
| Stau2 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human STAU2 (NP_0011578521). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.