Learn More
Invitrogen™ Staufen Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592142
Description
Immunogen sequence: QAVGVSQGHH TKDFTRAAPN PAKATVTAMI ARELLYGGTS PTAETILKNN ISSGHVPHGP LTRPSEQLDY LSRVQGFQVE YKDFPKNNKN EFVSLINCSS QPPLISHGIG KDVESCHDMA ALNILKLLSE LDQQSTEMPR TGNGPMSVCG RC; Positive Samples: SW480, BT-474, K-562, Mouse liver, Mouse brain, Rat liver, Rat brain; Cellular Location: Cytoplasm, Rough endoplasmic reticulum.
STAU1 is a member of a family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. The human STAU1 also contains a microtubule- binding domain similar to that of microtubule-associated protein 1B, and binds tubulin. STAU1 has been shown to be present in the cytoplasm in association with the rough endoplasmic reticulum (RER), implicating this protein in the transport of mRNA via the microtubule network to the RER. STAU1 is also known to interact with influenza ribonucleoproteins NS1, NP, and PA and is required for efficient viral replication.
Specifications
Staufen | |
Polyclonal | |
Unconjugated | |
Stau1 | |
5830401L18Rik; AW549911; C85792; Double-stranded RNA-binding protein Staufen homolog 1; PPP1R150; protein phosphatase 1, regulatory subunit 150; RP3-470L14.2; STAU; Stau1; staufen (RNA binding protein) homolog 1 (Drosophila); staufen double-stranded RNA binding protein 1; staufen RNA binding protein homolog 1; staufen, RNA binding protein, homolog 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20853, 6780, 84496 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95793, Q9Z108 | |
Stau1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 426-577 of human Staufen (NP_0593472). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.