Learn More
Invitrogen™ Staufen Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HepG2, Molt-4, Raji, Neuro2A, 293 (input), Stau1-IP (GTX106566), Post-IP lysate from control rabbit IgG-IP, Post-IP lysate from Stau1-IP. Predicted reactivity: Mouse (95%), Rat (94%), Zebrafish (83%), Xenopus laevis (86%), Chicken (94%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Staufen |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.93 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | Stau1 |
| Gene Accession No. | O95793, Q9Z108 |
| Gene Alias | 5830401L18Rik; AW549911; C85792; Double-stranded RNA-binding protein Staufen homolog 1; PPP1R150; protein phosphatase 1, regulatory subunit 150; RP3-470L14.2; STAU; Stau1; staufen (RNA binding protein) homolog 1 (Drosophila); staufen double-stranded RNA binding protein 1; staufen RNA binding protein homolog 1; staufen, RNA binding protein, homolog 1 |
| Gene Symbols | Stau1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.