Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staurosporine, 99+%, Thermo Scientific Chemicals

Catalog No. p-7042530
Click to view available options
Quantity:
1 mg
5 mg
10 mg

Staurosporine (staurosporin, CAS # 62996-74-1) is a bacterial alkyloid isolated from Streptomyces staurosporeus.

Broad spectrum protein kinase inhibitor Staurosporine is used as anti-fungal and anti-hypertensive, which results to study the anti-cancer treatment. It acts as a precursor to protein kinase inhibitor midostaurin. It is also used to induce cell death in Swiss Webster primary macrophage cells and to study the effect of caspase activation on human sperm quality.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Staurosporine is an indolocarbazole alkaloid capable of permeating cell membranes
  • Staurosporine has anti-cancer properties that can induce apoptosis
  • Staurosporine is a broad inhibitor of protein kinases, preventing growth and division cycles

Application

  • Staurosporine is used for research on acute myeloid leukemia
  • It is suitable for research applications including cell culture and biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
MDL Number MFCD00077402
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym Antibiotic AM-2282
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Type Staurosporine
Physical Form Powder
Melting Point 240°C to 243°C
Color Yellow
Quantity 1 mg
Beilstein 1060573
Merck Index 14,8802
Solubility Information Soluble in DMSO or ethanol;Soluble in dimethyl sulfoxide ,dimethyl formamide,ethyl acetate,hot acetone and ethanol; Slightly soluble in chloroform and methanol; Insoluble in water.
Formula Weight 466.54
Percent Purity ≥99%
Chemical Name or Material Staurosporine
Show More Show Less
Hazard Category H351
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P280-P308+P313-P501c
RTECSNumber KD5084000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.