Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Staurosporine, Fisher BioReagents
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 μg
500 μg
Packaging:
Poly Micro Tube
Description
Staurosporine is a potent protein kinase inhibitor useful for studies on protein phosphorylation in the regulation of cellular functions.

Specifications
Specifications
Melting Point | 270°C |
Color | White |
Assay Percent Range | > 98.0 % |
Packaging | Poly Micro Tube |
Quantity | 100 μg |
Merck Index | 15, 8928 |
Residual Solvents | Pass Test |
Formula Weight | 466.5 |
Percent Purity | >98.0% |
Physical Form | Solid |
Show More |
Safety and Handling

Hazard Statement | CAUTION! |
Precautionary Statement | Emergency Overview Limited evidence of a carcinogenic effect. May be harmful by inhalation, in contact with skin and if swallowed. Moisture sensitive. Sensitivity to light. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Use personal protective equipment. Do not breathe dust. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. NFPA Health:1 Flammability:1 Instability:1 |
Recommended Storage | 4°C, protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction