Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STBD1 Monoclonal Antibody (OTI6E9), TrueMAB™, OriGene
SDP

Catalog No. 501684497
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4497 Supplier OriGene Supplier No. TA812279
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May have the capability to bind to carbohydrates.

Specifications

Antigen STBD1
Applications Western Blot
Classification Monoclonal
Clone OTI6E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene STBD1
Gene Accession No. O95210
Gene Alias GENEX3414, GENX-3414
Gene Symbols STBD1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 24-358 of human STBD1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8987
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.