Learn More
STBD1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304481AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May have the capability to bind to carbohydrates.Specifications
STBD1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95210 | |
STBD1 | |
Between 75 and 125 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
STBD1 | |
GENEX3414, genethonin 1, starch-binding domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8987 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.