Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STC1 Monoclonal Antibody (C13)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA541678
This item is not returnable.
View return policy
Description
STC1 Monoclonal Antibody for Western Blot
This gene encodes a secreted, homodimeric glycoprotein that is expressed in a wide variety of tissues and may have autocrine or paracrine functions. The gene contains a 5' UTR rich in CAG trinucleotide repeats. The encoded protein contains 11 conserved cysteine residues and is phosphorylated by protein kinase C exclusively on its serine residues. The protein may play a role in the regulation of renal and intestinal calcium and phosphate transport, cell metabolism, or cellular calcium/phosphate homeostasis. Overexpression of human stanniocalcin 1 in mice produces high serum phosphate levels, dwarfism, and increased metabolic rate. This gene has altered expression in hepatocellular, ovarian, and breast cancers.
Specifications
STC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P52823 | |
STC1 | |
Recombinant protein Stanniocalcin 1. The antigen corresponds to amino acid range 28-247 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
C13 | |
Unconjugated | |
STC1 | |
stanniocalcin; stanniocalcin 1; stanniocalcin-1; STC; STC1; STC-1 | |
Mouse | |
Protein A/G | |
RUO | |
6781 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction