Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STC1 Monoclonal Antibody (C13)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543563
This item is not returnable.
View return policy
Description
STC1 Monoclonal Antibody for Western Blot
This gene encodes a secreted, homodimeric glycoprotein that is expressed in a wide variety of tissues and may have autocrine or paracrine functions. The gene contains a 5' UTR rich in CAG trinucleotide repeats. The encoded protein contains 11 conserved cysteine residues and is phosphorylated by protein kinase C exclusively on its serine residues. The protein may play a role in the regulation of renal and intestinal calcium and phosphate transport, cell metabolism, or cellular calcium/phosphate homeostasis. Overexpression of human stanniocalcin 1 in mice produces high serum phosphate levels, dwarfism, and increased metabolic rate. This gene has altered expression in hepatocellular, ovarian, and breast cancers.
Specifications
STC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P52823 | |
STC1 | |
Recombinant protein Stanniocalcin 1. The antigen corresponds to amino acid range 28-247 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
C13 | |
Unconjugated | |
STC1 | |
stanniocalcin; stanniocalcin 1; stanniocalcin-1; STC; STC1; STC-1 | |
Mouse | |
Protein A/G | |
RUO | |
6781 | |
-20°C | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction