Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STC2 Monoclonal Antibody (339CT13.2.6)
GREENER_CHOICE

Catalog No. PIMA537531
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537531 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537531 Supplier Invitrogen™ Supplier No. MA537531
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STC2 Monoclonal Antibody for Western Blot

Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STC2
Applications Western Blot
Classification Monoclonal
Clone 339CT13.2.6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STC2
Gene Accession No. O76061
Gene Alias AW125853; mustc2; stanniocalcin 2; stanniocalcin-2; Stanniocalcin-related protein; STC2; STC-2; Stc2l; STC-related protein; STCRP
Gene Symbols STC2
Host Species Mouse
Immunogen STC2 recombinant protein.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgA κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.