Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583770
Description
Immunogen sequence: SAIQKPPTAP PERQPQVDRT KLSRAHHGEA GHHLPEPSSR ETGRGAKGER GSKSHPNAHA RGRVGGLGAQ GPSGSSEWED EQSEYSDIRR.
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
Specifications
STC2 | |
Polyclonal | |
Unconjugated | |
STC2 | |
AW125853; mustc2; stanniocalcin 2; stanniocalcin-2; Stanniocalcin-related protein; STC2; STC-2; Stc2l; STC-related protein; STCRP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O76061 | |
STC2 | |
Recombinant protein corresponding to Human STC2. Recombinant protein control fragment (Product #RP-100450). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction