Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531946 Supplier Invitrogen™ Supplier No. PA531946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted reactivity: Mouse (80%), Rhesus Monkey (97%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STCH
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Hspa13
Gene Accession No. P48723
Gene Alias 1600002I10Rik; AV006182; B230217N24Rik; Heat shock 70 kDa protein 13; heat shock protein 13; heat shock protein 70 family, member 13; heat shock protein 70kDa family member 13; heat shock protein 70kDa family, member 13; heat shock protein family A (Hsp70) member 13; Hspa13; microsomal stress 70 protein ATPase core; microsomal stress-70 protein ATPase core; STCH; stress 70 protein chaperone microsome-associated 60 kDa protein; stress 70 protein chaperone microsome-associated 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kD; stress 70 protein chaperone, microsome-associated, 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kDa; stress 70 protein chaperone, microsome-associated, human homolog; stress-70 protein chaperone microsome-associated 60 kDa protein; testis secretory sperm-binding protein Li 199a
Gene Symbols Hspa13
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STCH. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6782
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.