Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592143 Supplier Invitrogen™ Supplier No. PA592143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPVANAVISV PAEFDLKQRN STIEAANLAG LKILRVINEP TAAAMAYGLH KADVFHVLVI DLGGGTLDVS LLNKQGGMFL TRAMSGNNKL GGQDFNQRLL QYLYKQIYQT YGFVPSRKEE IHRLRQAVEM VKLNLTLHQS AQLSVLLTVE EQDRKEPHSS DTELPKDKLS SADDHRVNSG FGRGLSDKKS GESQVLFETE ISRKLFDTLN EDLFQKILVP IQQVLKEGHL EKTEIDEVVL VGGSTRIPRI RQVIQEFFGK DPNTSVDPDL AVVTGVAIQA GIDGGSWPLQ VSALEIPNKH LQKTNFN; Positive Samples: U-87MG; Cellular Location: Endoplasmic reticulum, Microsome.

STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STCH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Hspa13
Gene Accession No. O35162, P48723, Q8BM72
Gene Alias 1600002I10Rik; AV006182; B230217N24Rik; Heat shock 70 kDa protein 13; heat shock protein 13; heat shock protein 70 family, member 13; heat shock protein 70kDa family member 13; heat shock protein 70kDa family, member 13; heat shock protein family A (Hsp70) member 13; Hspa13; microsomal stress 70 protein ATPase core; microsomal stress-70 protein ATPase core; STCH; stress 70 protein chaperone microsome-associated 60 kDa protein; stress 70 protein chaperone microsome-associated 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kD; stress 70 protein chaperone, microsome-associated, 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kDa; stress 70 protein chaperone, microsome-associated, human homolog; stress-70 protein chaperone microsome-associated 60 kDa protein; testis secretory sperm-binding protein Li 199a
Gene Symbols Hspa13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-471 of human HSPA13 (NP_0088793).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110920, 29734, 6782
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.