Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson Gloves Stealth Dog Fight Gloves

Catalog No. 17099795
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large

Stealth Dog Fight Gloves have a HPPE/glass/steel/nylon shell with ANSI A5 cut resistance for superior protection.

  • HPPE/glass/steel/nylon shell with ANSI A5 cut resistance
  • Heavy duty TPR on fingers, thumb, knuckles, and back of hand
  • Hi-vis sandy nitrile finish
  • Extended snug-fitting knit wrist

Specifications

Certifications/Compliance ANSI CUT, ANSI PUNCTURE, ANSI IMPACT
Certifications/Compliance (Europe) EN388
Coated Area Palm
Construction Seamless Knit
Cuff Material HPPE, Glass, Steel, Nylon
Coating Material Nitrile
Color Hi-vis Yellow, Gray
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material HPPE, Glass, Steel Fiber, Nylon, Sandy Nitrile
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN388:4X42EP
For Use With (Application) Industrial, Warehouse, Sheet Metal, Construction, General Labor
Gender Men's
Glove Cuff Color Gray
Glove Cuff Style Continuous Knit
Liner Material Unlined
Lining Unlined
Model 357TPR
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Stealth
Length (English) 9.5 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.