Learn More
Cancer Diagnostics, Inc.™ Stealth Safety Scalpels
SDP
Catalog No. p-4272587
Click to view available options
Size:
# 22
# 60
  • Integrated rigid plastic handle with protective sheath.
  • Slide sheath down to use, slide up when not in use.
  • Dispose when dull.
Sizes:
#22 and #60 and #70 (round tip)
Specifications
Product Type Finger Scalpel
Size # 60
Disposable Single-use
Sterility Non-sterile
Includes Handle
Handle Material Plastic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.