Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson Gloves Stealth Stinger Cut A2 Gloves

Catalog No. 17099964
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large

Stealth Stinger Cut A2 Gloves have a 15 gg fine gauge HPPE, nylon, glass, and spandex shell that is cut resistant offering superior protection and durability while providing maximum dexterity.

  • 15 gg Fine gauge HPPE, nylon, glass, and spandex shell
  • Lightweight polyurethane coated palm for a sure-grip
  • Snug-fitting knit wrist

Specifications

Certifications/Compliance ANSI Cut, ANSI Puncture
Certifications/Compliance (Europe) EN388
Coated Area Palm
Construction Seamless Knit
Coating Material Polyurethane
Color Blue, Black
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Latex Free Yes
Material Nylon, Spandex, Polyurethane
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN388:3X42B
For Use With (Application) Light Industrial, General Labor, High dexterity
Gender Men's
Glove Cuff Style Elasticated
Liner Material Unlined
Lining Lined
Model 359
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Stealth
Yarn Gauge 15
Length (English) 9.5 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.