Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STEAP1 Antibody (J2D2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP107094IR
Description
STEAP1 Monoclonal specifically detects STEAP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
STEAP1 | |
Monoclonal | |
DyLight 755 | |
NP_036581 | |
STEAP1 | |
Recombinant human STEAP1 (1-70aa) purified from E. coli (NP_036581). | |
0.1 mL | |
Cancer | |
26872 | |
Human, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
J2D2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 1.16.1.-, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
STEAP1 (J2D2) | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction