Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STEAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111007
Description
Immunogen sequence: CLRKKILKIR HGWEDVTKIN KTEICSQL
STEAP1 is predominantly expressed in prostate tissue, and is found to be upregulated in multiple cancer cell lines. The gene product is predicted to be a six-transmembrane protein, and was shown to be a cell surface antigen significantly expressed at cell-cell junctions.
Specifications
STEAP1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHE8 | |
STEAP1 | |
Recombinant Protein corresonding to Human STEAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
STEAP1 | |
2410007B19Rik; Metalloreductase STEAP1; PRSS24; Six-transmembrane epithelial antigen of prostate 1; Steap; STEAP1; STEAP-1; tcag7.1170 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26872 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction