Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STEAP1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568752
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 77%; Rabbit: 93%; Rat: 93%.
STEAP1B is a protein coding gene.
Specifications
STEAP1B | |
Polyclonal | |
Unconjugated | |
STEAP1B | |
STEAP family member 1B; STEAP family protein MGC87042; STEAP1B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
256227 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6NZ63 | |
STEAP1B | |
synthetic peptide directed towards the N-terminal region of human STEAP1B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction