Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STEAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533060
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533060 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533060 Supplier Invitrogen™ Supplier No. PA533060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Baboon, Monkey, Galago, Marmoset, Mouse, Rat, Hamster, Elephant, Panda, Bat, Dog, Rabbit, Horse, Opossum, Guinea pig, Armadillo, Platypus (100%); Bovine, Pig, Turkey, Chicken, Zebrafish (93%); Shrew, Zebra finch, Xenopus (87%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

The six-transmembrane epithelial antigen of prostate 2 (STEAP2) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 3 and STEAP4), STEAP2 promotes both iron and copper reduction. STEAP2 expression in transfected cells also correlated with iron or copper uptake, suggesting that the STEAP family of proteins may function to stimulate iron and copper uptake. STEAP2 is widely expressed in many tissues in the plasma membrane, but is most highly expressed in prostate. At least three isoforms of STEAP2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene STEAP2
Gene Accession No. Q8BWB6, Q8NFT2
Gene Alias 4921538B17Rik; AI930049; AW045895; IPCA1; IPCA-1; Metalloreductase STEAP2; PCANAP1; prostate cancer associated protein 1; prostate cancer-associated protein 1; protein upregulated in metastatic prostate cancer; Protein up-regulated in metastatic prostate cancer; PUMPCn; six transmembrane epithelial antigen of prostate 2; six transmembrane epithelial antigen of the prostate 2; six-transmembrane epithelial antigen of prostate 2; SixTransMembrane protein of prostate 1; STAMP1; STEAP family member 2, metalloreductase; Steap2; STEAP2 metalloreductase; STMP; UNQ6507/PRO23203
Gene Symbols STEAP2
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from internal region of human STEAP2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050610, 100328631, 100729189, 100768345, 261729, 312052, 482291, 74051
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Hamster, Horse, Guinea Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.