Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STEAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587478 Supplier Invitrogen™ Supplier No. PA587478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPEEMDKPLI SLHLVDSDSS LAKVPDEAPK VGILGSGDFA RSLATRLVGS GFKVVVGSRN PKRTARLFPS AAQVTFQEEA VSSPEVIFVA VFREHYSSLC SLSDQLAGKI LVDVSNPTEQ EHLQHRESNA EYLASLFPTC TVVKAFNVIS AWTLQAGPRD GNRQVPICGD QPEAKRAVSE; Positive Samples: Mouse heart; Cellular Location: Endosome membrane, Multi-pass membrane protein.

The six-transmembrane epithelial antigen of prostate 3 (STEAP3) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 2 and STEAP4), STEAP3 promotes both iron and copper reduction. STEAP3 is highly expressed in hematopoietic tissues and colocalizes with the transferrin endosome. Overexpression of STEAP3 stimulates iron reduction; mice lacking STEAP3 are deficient in erythroid ferrireductase activity, suggesting that STEAP3 is an endosomal ferrireductase required for transferrin-dependent iron uptake in erythroid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STEAP3
Gene Accession No. Q8CI59
Gene Alias 1010001D01Rik; AHMIO2; dudlin 2; dudlin-2; dudulin 2; dudulin-2; hpHyde; hTSAP6; Metalloreductase STEAP3; pHyde; Protein nm1054; six-transmembrane epithelial antigen of prostate 3; STEAP family member 3; STEAP family member 3, metalloreductase; STEAP3; STEAP3 metalloreductase; STMP3; TSAP6; tumor suppressor pHyde; tumor suppressor-activated pathway protein 6
Gene Symbols STEAP3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68428
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.