Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STEAP3/TSAP6 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176824F
Description
STEAP3/TSAP6 Polyclonal antibody specifically detects STEAP3/TSAP6 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
STEAP3/TSAP6 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FITC | |
dudlin 2, dudlin-2, dudulin 2, dudulin-2, Dudulin-2,1010001D01Rik, EC 1.16.1.-, hpHyde, hTSAP6, metalloreductase STEAP3, pHyde, six transmembrane prostate protein 3, Six-transmembrane epithelial antigen of prostate 3, STEAP family member 3, TSAP6STMP3, tumor suppressor pHyde, Tumor suppressor-activated pathway protein 6 | |
Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human STEAP3. The immunogen is located within the first 50 amino acids of STEAP3. Amino Acid Squence: DSDSSLAKVPDEAPK | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
55240 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction