Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STEAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534225
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534225 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534225 Supplier Invitrogen™ Supplier No. PA534225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Monkey, Marmoset (93%) Gibbon, Mouse, Bat, Horse (87%) Hamster, Elephant, Panda, Rabbit, Pig, Xenopus (80%).

The six-transmembrane epithelial antigen of prostate 4 (STEAP4) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 2 and STEAP3), STEAP4 promotes both iron and copper reduction. STEAP4 is highly expressed in placenta, lung, heart and prostate tissues, and overexpressed in prostate cancer cells compared to normal prostate cells. Overexpression of STEAP4 in prostate cells significantly increases cell growth and colony formation, suggesting STEAP4 may have a role in cell proliferation and prostate cancer progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEAP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene STEAP4
Gene Accession No. Q687X5
Gene Alias 1110021O17Rik; Ab1-219; AI481214; DKFZp666D049; dudulin 4; dudulin-4; FLJ23153; Metalloreductase STEAP4; six transmembrane prostate protein 2; six-transmembrane epithelial antigen of prostate 4; SixTransMembrane protein of prostate 2; STAMP2; STEAP family member 4; STEAP4; STEAP4 metalloreductase; TIARP; Tnfa-induced adipose-related protein; Tnfaip9; Tumor necrosis factor, alpha-induced protein 9; tumor necrosis factor-alpha-induced adipose-related protein; tumor necrosis-alpha-induced adipose-related protein
Gene Symbols STEAP4
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-Terminus of human STEAP4.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.