Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearamide 90.0+%, TCI America™
SDP

Catalog No. S0075500G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g
500 g

Chemical Identifiers

CAS 124-26-5
Molecular Formula C18H37NO
Molecular Weight (g/mol) 283.50
MDL Number MFCD00008038
InChI Key LYRFLYHAGKPMFH-UHFFFAOYSA-N
Synonym stearamide, stearylamide, octadecylamide, stearoylamide, stearoylamine, octadecamide, stearic acid amide, stearic amide, kemamide s, adogen 42
PubChem CID 31292
ChEBI CHEBI:34900
IUPAC Name octadecanamide
SMILES CCCCCCCCCCCCCCCCCC(N)=O

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 500 g
Formula Weight 283.50
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stearamide
EINECSNumber (2)-0824
RTECSNumber RG0182000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.