Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearic acid, 98%

Catalog No. p-3346532
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 57-11-4 | C18H36O2 | 284.48 g/mol

A saturated fatty acid Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A saturated fatty acid Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

Solubility
Soluble in ethanol, dimethyl sulfoxide, dimethyl formamide, carbon terachloride, toluene, chloroform, acetone and carbondisulfide. Insoluble in water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-11-4
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
Synonym stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
PubChem CID 5281
ChEBI CHEBI:28842
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 69°C to 72°C
Density 0.941
Boiling Point 232°C (15 mm)
Flash Point 196°C (384°F)
Assay Percent Range 98%
Quantity 5 g
Beilstein 608585
Solubility Information Soluble in ethanol,dimethyl sulfoxide,dimethyl formamide,carbon terachloride,toluene,chloroform,acetone and carbondisulfide. Insoluble in water.
Formula Weight 284.48
Chemical Name or Material Stearic Acid
Hazard Category H500
EINECSNumber 200-313-4
RTECSNumber WI2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.